Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Deoxy-D-Galactose, MP Biomedicals™

Catalog No. ICN15080001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15080001 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15080001 Supplier MP Biomedicals, Inc Supplier No. 0215080001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical reagent used as an inhibitor of fucosylation.

Inhibitor of fucosylation
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1949-89-9
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00014649
InChI Key PMMURAAUARKVCB-ONLQHTGVNA-N
Synonym 2-deoxy-d-galactose, 3r,4r,5r-3,4,5,6-tetrahydroxyhexanal, d-lyxo-hexose, 2-deoxy, unii-531k2iok5q, 2-deoxy-d-lyxo-hexose, lyxo-hexose, 2-deoxy, ksc179c4l
PubChem CID 102191
ChEBI CHEBI:27411
IUPAC Name (4R,5R,6R)-6-(hydroxymethyl)oxane-2,4,5-triol
SMILES OC[C@H]1OC(O)C[C@@H](O)[C@H]1O

Specifications

Quantity 1 g
Formula Weight 164.2
Grade Research
Physical Form Solid
Chemical Name or Material 2-Deoxy-D-Galactose
Recommended Storage Store at +4°C.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.