accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNOO

CAS RN 19499-61-7

IUPAC Name: methyl[(3-nitrophenyl)methyl]amine
Synonyms: methyl[(3-nitrophenyl)methyl]amine
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: NTPAPKLGADEFAM-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results