Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-N-(3-nitrobenzyl)amine, 97+%, Thermo Scientific™

Catalog No. MO00890DE
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO00890DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO00890DE Supplier Thermo Scientific Chemicals Supplier No. MO00890DE
Add to Cart

Chemical Identifiers

CAS 19499-61-7
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
InChI Key NTPAPKLGADEFAM-UHFFFAOYSA-N
Synonym n-methyl-1-3-nitrophenyl methanamine, n-methyl-n-3-nitrobenzyl amine, methyl 3-nitrophenyl methyl amine, benzenemethanamine, n-methyl-3-nitro, 3-nitro-n-methylbenzylamine, n-methyl-3-nitrobenzylamine, n-3-nitrophenylmethyl methylamine, n-methyl-1-3-nitrophenyl methylamine
PubChem CID 409439
SMILES CNCC1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 166.18
Percent Purity 95%
Chemical Name or Material N-Methyl-N-(3-nitrobenzyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.