accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19549-98-5

IUPAC Name: 3,6-dimethylhept-1-yn-3-ol
Synonyms: 3,6-dimethylhept-1-yn-3-ol
Molecular Weight (g/mol): 140.23
Molecular Formula: C9H16O
InChi Key: COWFALGRNJAHKW-UHFFFAOYNA-N
SMILES: CC(C)CCC(C)(O)C#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results