accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3HOH3C

CAS RN 19550-08-4

IUPAC Name: 3,4-dimethylhexan-3-ol
Synonyms: 3,4-dimethylhexan-3-ol
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: FJXOYCIYKQJAAF-UHFFFAOYNA-N
SMILES: CCC(C)C(C)(O)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results