CAS RN 1956-10-1
IUPAC Name:
4-nitrophenyl octanoate
Synonyms:
4-nitrophenyl octanoate
Molecular Weight (g/mol):
265.31
Molecular Formula:
C14H19NO4
InChi Key:
GGIDEJQGAZSTES-UHFFFAOYSA-N
SMILES:
CCCCCCCC(=O)OC1=CC=C(C=C1)[N+]([O-])=O
1
–
2
of
2
results
1
Promotions Available
4-Nitrophenyl octanoate, 96%
4-Nitrophenyl octanoate, 96%
CAS: 1956-10-1 Molecular Formula: C14H19NO4 Molecular Weight (g/mol): 265.31 MDL Number: MFCD00024665 InChI Key: GGIDEJQGAZSTES-UHFFFAOYSA-N
2
R&D Systems™ Galanthamine hydrobromide
CAS: 1956-10-1 Molecular Formula: C14H19NO4