accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19653-33-9

IUPAC Name: ethyl 3-(piperidin-1-yl)propanoate
Synonyms: ethyl 3-(piperidin-1-yl)propanoate
Molecular Weight (g/mol): 185.27
Molecular Formula: C10H19NO2
InChi Key: MPAWVDTVWPPKJQ-UHFFFAOYSA-N
SMILES: CCOC(=O)CCN1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results