accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNHN

CAS RN 1968-05-4

IUPAC Name: 3-[(1H-indol-3-yl)methyl]-1H-indole
Synonyms: 3,3'-diindolylmethane 3,3'-diindolymethane DIM
Molecular Weight (g/mol): 246.31
Molecular Formula: C17H14N2
InChi Key: VFTRKSBEFQDZKX-UHFFFAOYSA-N
SMILES: C(C1=CNC2=CC=CC=C12)C1=CNC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results