accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOO

CAS RN 19693-75-5

IUPAC Name: 2-methoxy-1,3-dioxolane
Synonyms: 2-methoxy-1,3-dioxolane
Molecular Weight (g/mol): 104.11
Molecular Formula: C4H8O3
InChi Key: VRAYTNFBRROPJU-UHFFFAOYSA-N
SMILES: COC1OCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results