Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-1,3-dioxolane 98.0+%, TCI America™
SDP

Catalog No. M081625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M081625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M081625G Supplier TCI America Supplier No. M081625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19693-75-5
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.105
MDL Number MFCD00003208
InChI Key VRAYTNFBRROPJU-UHFFFAOYSA-N
PubChem CID 29744
IUPAC Name 2-methoxy-1,3-dioxolane
SMILES COC1OCCO1

Specifications

Color Colorless
Boiling Point 130°C
Quantity 25 g
UN Number 1993
Formula Weight 104.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxy-1,3-dioxolane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.