accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19718-45-7

IUPAC Name: 2-propoxynaphthalene
Synonyms: 2-propoxynaphthalene
Molecular Weight (g/mol): 186.25
Molecular Formula: C13H14O
InChi Key: UEXFDEKTELCPNF-UHFFFAOYSA-N
SMILES: CCCOC1=CC=C2C=CC=CC2=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results