Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propoxynaphthalene, TCI America™
SDP

Catalog No. N043225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N043225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N043225G Supplier TCI America Supplier No. N043225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19718-45-7
Molecular Formula C13H14O
Molecular Weight (g/mol) 186.25
MDL Number MFCD00021610
InChI Key UEXFDEKTELCPNF-UHFFFAOYSA-N
Synonym 2-Naphthyl Propyl Ether
PubChem CID 243587
IUPAC Name 2-propoxynaphthalene
SMILES CCCOC1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 40°C
Boiling Point 305°C
Quantity 25 g
Formula Weight 186.25
Physical Form Crystalline Powder
Chemical Name or Material 2-Propoxynaphthalene
EINECSNumber (4)-0362
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.