accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19718-92-4

IUPAC Name: (4,4-dimethoxybutyl)dimethylamine
Synonyms: (4,4-dimethoxybutyl)dimethylamine
Molecular Weight (g/mol): 161.25
Molecular Formula: C8H19NO2
InChi Key: WDZKKBDOGYBYBG-UHFFFAOYSA-N
SMILES: COC(CCCN(C)C)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results