accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19727-83-4

IUPAC Name: 6-nitro-2,3-dihydro-1H-indole
Synonyms: 6-nitro-2,3-dihydro-1H-indole
Molecular Weight (g/mol): 164.16
Molecular Formula: C8H8N2O2
InChi Key: LTNYDSMDSLOMSM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2CCNC2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)