Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Nitroindoline, 98+%, Thermo Scientific™

Catalog No. AAB2196106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2196106 5 g
AAB2196103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2196106 Supplier Thermo Scientific Chemicals Supplier No. B2196106

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19727-83-4
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.16
MDL Number MFCD00005710
InChI Key LTNYDSMDSLOMSM-UHFFFAOYSA-N
Synonym 6-nitroindoline, indoline, 6-nitro, 1h-indole, 2,3-dihydro-6-nitro, 6-nitro-indoline, 2,3-dihydro-6-nitro-1h-indole, ccris 3302, zlchem 357, pubchem7443
PubChem CID 29757
IUPAC Name 6-nitro-2,3-dihydro-1H-indole
SMILES [O-][N+](=O)C1=CC=C2CCNC2=C1

Specifications

Melting Point 66°C to 70°C
Quantity 5 g
Beilstein 156434
Formula Weight 164.16
Percent Purity ≥98%
Chemical Name or Material 6-Nitroindoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H341-H332
Toxic if swallowed.
Toxic in contact with skin.
Suspected of causing genetic defects.
Harmful if inhaled.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 243-257-6
RTECSNumber NM1950000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.