Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Nitroindoline 97.0+%, TCI America™
SDP

Catalog No. N04035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04035G 5 g
N04031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N04035G Supplier TCI America Supplier No. N04035G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19727-83-4
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.16
MDL Number MFCD00005710
InChI Key LTNYDSMDSLOMSM-UHFFFAOYSA-N
Synonym 6-nitroindoline, indoline, 6-nitro, 1h-indole, 2,3-dihydro-6-nitro, 6-nitro-indoline, 2,3-dihydro-6-nitro-1h-indole, ccris 3302, zlchem 357, pubchem7443
PubChem CID 29757
IUPAC Name 6-nitro-2,3-dihydro-1H-indole
SMILES [O-][N+](=O)C1=CC=C2CCNC2=C1

Specifications

Melting Point 67°C
Quantity 5 g
Formula Weight 164.16
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Nitroindoline
RTECSNumber NM1950000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.