accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19757-97-2

IUPAC Name: methyl 2-(hydroxymethoxy)acetate
Synonyms: methyl 2-(hydroxymethoxy)acetate
Molecular Weight (g/mol): 120.1
Molecular Formula: C4H8O4
InChi Key: RNVVYVPDHMKDTL-UHFFFAOYSA-N
SMILES: COC(=O)COCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results