accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3COOIONCH3CH3I

CAS RN 19774-82-4

IUPAC Name: hydrogen {2-[4-(2-butyl-1-benzofuran-3-carbonyl)-2,6-diiodophenoxy]ethyl}diethylamine chloride
Synonyms: hydrogen amiodarone chloride
Molecular Weight (g/mol): 681.78
Molecular Formula: C25H30ClI2NO3
InChi Key: ITPDYQOUSLNIHG-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCCCC1=C(C(=O)C2=CC(I)=C(OCCN(CC)CC)C(I)=C2)C2=CC=CC=C2O1
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)