Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amiodarone hydrochloride, R&D Systems™
SDP

Catalog No. 409550 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-955-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 40-955-0 Supplier R&D Systems Supplier No. 4095/50

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Broad spectrum ion channel blocker; antiarrhythmic

Amiodarone hydrochloride is a broad-spectrum ion channel blocker; blocks late INa, ICa, IKr and IKs and increases QT. Displays class III antiarrhythmic properties. Also exhibits fungicidal activity; elicits Ca2+ influx in Saccharomyces cerevisiae and causes mitochondrial fragmentation and cell death. Thought to stimulate autophagy by targeting upstream mTORC1 control pathways. Selectively toxic to NSCs in hESC-derived cell populations. Amiodarone depolarizes the resting membrane potential of the triple-negative breast cancer (TNBC) cells and decreases cell migration.

Chemical Identifiers

CAS 19774-82-4
Molecular Formula C25H30ClI2NO3
Molecular Weight (g/mol) 681.778
InChI Key ITPDYQOUSLNIHG-UHFFFAOYSA-N
Synonym amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron
PubChem CID 441325
IUPAC Name (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride
SMILES CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl

Specifications

Quantity 50 mg
Formula Weight 681.77
Percent Purity >99%
Product Line Tocris
Chemical Name or Material Amiodarone hydrochloride
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.