accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHNOH(E/Z)(E/Z)(E/Z)(E/Z)

CAS RN 198022-70-7

IUPAC Name: N-(4-hydroxyphenyl)icosa-5,8,11,14-tetraenamide
Synonyms: N-(4-hydroxyphenyl)icosa-5,8,11,14-tetraenamide
Molecular Weight (g/mol): 395.59
Molecular Formula: C26H37NO2
InChi Key: IJBZOOZRAXHERC-UHFFFAOYSA-N
SMILES: CCCCCC=CCC=CCC=CCC=CCCCC(=O)NC1=CC=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)