CAS RN 19889-77-1
IUPAC Name:
piperidine-2-carboxamide
Synonyms:
piperidine-2-carboxamide
Molecular Weight (g/mol):
128.18
Molecular Formula:
C6H12N2O
InChi Key:
XIMBESZRBTVIOD-UHFFFAOYNA-N
SMILES:
NC(=O)C1CCCCN1
1
–
1
of
1
results
1
2-Piperidinecarboxamide 98.0+%, TCI America™
CAS: 19889-77-1 Molecular Formula: C6H12N2O Molecular Weight (g/mol): 128.18 MDL Number: MFCD05663784 InChI Key: XIMBESZRBTVIOD-UHFFFAOYNA-N PubChem CID: 140623 IUPAC Name: piperidine-2-carboxamide SMILES: NC(=O)C1CCCCN1