accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3C(S)OH(R)

CAS RN 19894-97-4

IUPAC Name: [(1R,5S)-6,6-dimethylbicyclo[3.1.1]hept-2-en-2-yl]methanol
Synonyms: [(1R,5S)-6,6-dimethylbicyclo[3.1.1]hept-2-en-2-yl]methanol
Molecular Weight (g/mol): 152.24
Molecular Formula: C10H16O
InChi Key: RXBQNMWIQKOSCS-IUCAKERBSA-N
SMILES: CC1(C)[C@@H]2C[C@H]1C(CO)=CC2
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results