accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOHOHOHOHO

CAS RN 1990-29-0

IUPAC Name: 2,3,4,5,6-pentahydroxyhexanal
Synonyms: dextrose corn sugar (14C)-glucose D(+)-glucose (U-14C)glucose (14C)D-glucose sirup D-(U-14C)glucose glucose (D) D(+)-mannose
Molecular Weight (g/mol): 180.16
Molecular Formula: C6H12O6
InChi Key: GZCGUPFRVQAUEE-UHFFFAOYNA-N
SMILES: OCC(O)C(O)C(O)C(O)C=O
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)