Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Altrose, MP Biomedicals™

Catalog No. ICN19120825 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19120825 25 mg
ICN19120810 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19120825 Supplier MP Biomedicals, Inc Supplier No. 0219120825
Add to Cart
Add to Cart

Used as a substrate to identify, differentiate and characterize aldose isomerases such as L-fucose isomerase from Caldicellulosiruptor saccharolyticus and d-Arabinose isomerase (d-AI) from Bacillus pallidus (B. pallidus) and Klebsiella pneumoniae

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1990-29-0
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
InChI Key GZCGUPFRVQAUEE-ARQDHWQXSA-N
Synonym d-altro-hexose, 2s,3r,4r,5r-2,3,4,5,6-pentahydroxyhexanal, unii-5pln1o36ff, 5pln1o36ff, d-+-altrose, aldehydo-d-altrose
PubChem CID 94780
ChEBI CHEBI:28385
IUPAC Name (2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Color White
Quantity 25 mg
Formula Weight 180.16
Physical Form Powder
Chemical Name or Material D-(+)-Altrose
EINECSNumber 217-870-4
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.