CAS RN 1990509-31-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-(Triallylsilyl)propyl Methacrylate (stabilized with MEHQ) 92.0+%, TCI America™
CAS: 1990509-31-3 Molecular Formula: C16H26O2Si Synonym: Methacrylic Acid 3-(Triallylsilyl)propyl Ester