accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OHONHSOOBr

CAS RN 199850-67-4

IUPAC Name: 2-{4'-bromo-[1,1'-biphenyl]-4-sulfonamido}-3-methylbutanoic acid
Synonyms: 2-{4'-bromo-[1,1'-biphenyl]-4-sulfonamido}-3-methylbutanoic acid
Molecular Weight (g/mol): 412.3
Molecular Formula: C17H18BrNO4S
InChi Key: GJOCABIDMCKCEG-UHFFFAOYNA-N
SMILES: CC(C)C(NS(=O)(=O)C1=CC=C(C=C1)C1=CC=C(Br)C=C1)C(O)=O
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results