CAS RN 199864-86-3
IUPAC Name:
4-(4-fluoronaphthalen-1-yl)-6-(propan-2-yl)pyrimidin-2-amine hydrochloride
Synonyms:
4-(4-fluoronaphthalen-1-yl)-6-isopropylpyrimidin-2-amine hydrochloride
Molecular Weight (g/mol):
317.79
Molecular Formula:
C17H17ClFN3
InChi Key:
MKJPYBJBPRFMHL-UHFFFAOYSA-N
SMILES:
Cl.CC(C)C1=NC(N)=NC(=C1)C1=C2C=CC=CC2=C(F)C=C1
1
–
1
of
1
results
1
R&D Systems™ RS 127445 hydrochloride
CAS: 199864-86-3 Molecular Formula: C17H17ClFN3