accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNCH2NNHBrBr

CAS RN 199934-16-2

IUPAC Name: 1λ⁵,9λ⁵,17,24-tetraazaheptacyclo[23.6.2.2⁹,¹⁶.2¹⁹,²².1³,⁷.0¹⁰,¹⁵.0²⁶,³¹]octatriaconta-1(31),3(38),4,6,9,11,13,15,19,21,25,27,29,32,34,36-hexadecaene-1,9-bis(ylium) dibromide
Synonyms: 1λ⁵,9λ⁵,17,24-tetraazaheptacyclo[23.6.2.2⁹,¹⁶.2¹⁹,²².1³,⁷.0¹⁰,¹⁵.0²⁶,³¹]octatriaconta-1(31),3(38),4,6,9,11,13,15,19,21,25,27,29,32,34,36-hexadecaene-1,9-bis(ylium) dibromide
Molecular Weight (g/mol): 654.45
Molecular Formula: C34H30Br2N4
InChi Key: KPNMQIKQVCWNTP-UHFFFAOYSA-N
SMILES: [Br-].[Br-].C1NC2=C3C=CC=CC3=[N+](CC3=CC(C[N+]4=C5C=CC=CC5=C(NCC5=CC=C1C=C5)C=C4)=CC=C3)C=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)