accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOO

CAS RN 200698-05-1

IUPAC Name: 9-benzyl-9H-carbazole-3,6-dicarbaldehyde
Synonyms: 9-benzylcarbazole-3,6-dicarbaldehyde
Molecular Weight (g/mol): 313.36
Molecular Formula: C21H15NO2
InChi Key: RZSXZJXJVWYRIU-UHFFFAOYSA-N
SMILES: O=CC1=CC2=C(C=C1)N(CC1=CC=CC=C1)C1=C2C=C(C=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results