accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFClH2NFFF

CAS RN 201593-90-0

IUPAC Name: 2-chloro-3,5-bis(trifluoromethyl)aniline
Synonyms: 2-chloro-3,5-bis(trifluoromethyl)aniline
Molecular Weight (g/mol): 263.57
Molecular Formula: C8H4ClF6N
InChi Key: JKUFETFGEJPHEM-UHFFFAOYSA-N
SMILES: NC1=CC(=CC(=C1Cl)C(F)(F)F)C(F)(F)F
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results