CAS RN 2022961-17-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Raphin 1
Inhibitor of the regulatory subunit PPP1R15B of protein phosphatase 1 (Kd = 33 nM); orally bioavailable and brain penetrant