accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOONOO

CAS RN 20261-31-8

IUPAC Name: 2-hydroxy-3-nitro-4H-chromen-4-one
Synonyms: 2-hydroxy-3-nitrochromen-4-one
Molecular Weight (g/mol): 207.14
Molecular Formula: C9H5NO5
InChi Key: SLXZSRMEGWRHRH-UHFFFAOYSA-N
SMILES: OC1=C(C(=O)C2=CC=CC=C2O1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results