Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-3-nitrocoumarin 98.0+%, TCI America™
SDP

Catalog No. H145825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H145825G 25 g
H14585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H145825G Supplier TCI America Supplier No. H145825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20261-31-8
Molecular Formula C9H5NO5
Molecular Weight (g/mol) 207.141
MDL Number MFCD00051671
InChI Key NZQAQAUWFHMVEM-UHFFFAOYSA-N
Synonym 4-hydroxy-3-nitrocoumarin, 4-hydroxy-3-nitro-2h-chromen-2-one, 3-nitro-4-hydroxycoumarin, 2h-1-benzopyran-2-one, 4-hydroxy-3-nitro, 2-hydroxy-3-nitrochromen-4-one, 4-hydroxy-3-nitro-chromen-2-one, 2h-1-benzopyran-2-one,4-hydroxy-3-nitro, 4-hydroxy-3-hydroxy oxido amino-2h-chromen-2-one, acmc-20amh1, maybridge1_005221
PubChem CID 54682640
IUPAC Name 4-hydroxy-3-nitrochromen-2-one
SMILES C1=CC=C2C(=C1)C(=C(C(=O)O2)[N+](=O)[O-])O

Specifications

Melting Point 171°C
Color Yellow
Quantity 25 g
Formula Weight 207.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-3-nitrocoumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.