CAS RN 202796-42-7
IUPAC Name:
1-[(cyclooct-1-en-1-yl)methyl]-4-(2,7-dichloro-9H-xanthene-9-amido)-1-ethylpiperidin-1-ium iodide
Synonyms:
1-(cyclooct-1-en-1-ylmethyl)-4-(2,7-dichloro-9H-xanthene-9-amido)-1-ethylpiperidin-1-ium iodide
Molecular Weight (g/mol):
655.44
Molecular Formula:
C30H37Cl2IN2O2
InChi Key:
FOAFBMYSXIGAOX-UHFFFAOYSA-N
SMILES:
[I-].CC[N+]1(CC2=CCCCCCC2)CCC(CC1)NC(=O)C1C2=C(OC3=C1C=C(Cl)C=C3)C=CC(Cl)=C2
1
–
1
of
1
results
1
Tocris Bioscience™ UCB 35625
CAS: 202796-42-7 Molecular Formula: C30H37Cl2IN2O2