accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNOCH3O

CAS RN 20306-75-6

IUPAC Name: N-methyl-3-oxobutanamide
Synonyms: N-methyl-3-oxobutanamide
Molecular Weight (g/mol): 115.13
Molecular Formula: C5H9NO2
InChi Key: ATWLCPHWYPSRBQ-UHFFFAOYSA-N
SMILES: CNC(=O)CC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results