accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2032-34-0

IUPAC Name: 3,3-diethoxypropanenitrile
Synonyms: 3,3-diethoxypropanenitrile
Molecular Weight (g/mol): 143.19
Molecular Formula: C7H13NO2
InChi Key: WBOXEOCWOCJQNK-UHFFFAOYSA-N
SMILES: CCOC(CC#N)OCC
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)