Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Diethoxy-2-cyanoethane 95%, Thermo Scientific™

Catalog No. AC269660250
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC269660250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC269660250 Supplier Thermo Scientific Chemicals Supplier No. 269660250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2032-34-0
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.19
MDL Number MFCD00040558
InChI Key WBOXEOCWOCJQNK-UHFFFAOYSA-N
Synonym 3,3-diethoxypropionitrile, 3,3-diethoxypropiononitrile, propanenitrile, 3,3-diethoxy, cyanoacetaldehyde diethyl acetal, 1,1-diethoxy-2-cyanoethane, 3,3-diethoxy propionitrile, cyadea, acmc-1cmza, 3,3-diethoxypropanitrile, 3,3-diethoxypropiononitril
PubChem CID 74851
IUPAC Name 3,3-diethoxypropanenitrile
SMILES CCOC(CC#N)OCC

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Brown to Yellow
Density 0.95
Infrared Spectrum Authentic
Refractive Index 1.4140 to 1.4160 (20°C, 589nm)
Linear Formula (C2H5O)2CHCH2CN
Quantity 25 mL
Formula Weight 143.19
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material 1, 1-Diethoxy-2-cyanoethane, 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.