CAS RN 2035509-96-5
IUPAC Name:
5-({2-ethoxy-4-[4-(4-methylpiperazin-1-yl)piperidine-1-carbonyl]phenyl}amino)-2-methanesulfonyl-9-methyl-2,4,6,9-tetraazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one
Synonyms:
5-({2-ethoxy-4-[4-(4-methylpiperazin-1-yl)piperidine-1-carbonyl]phenyl}amino)-2-methanesulfonyl-9-methyl-2,4,6,9-tetraazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one
Molecular Weight (g/mol):
648.78
Molecular Formula:
C32H40N8O5S
InChi Key:
HTFNVAVTYILUCF-UHFFFAOYSA-N
SMILES:
CCOC1=C(NC2=NC=C3N(C)C(=O)C4=CC=CC=C4N(C3=N2)S(C)(=O)=O)C=CC(=C1)C(=O)N1CCC(CC1)N1CCN(C)CC1
1
–
1
of
1
results
1
Tocris Bioscience™ AX 15836, > 98%
CAS: 2035509-96-5 Molecular Formula: C32H40N8O5S