accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaOSOO(E)

CAS RN 2039-44-3

IUPAC Name: sodium (1E)-2-phenylethene-1-sulfonate
Synonyms: sodium (1E)-2-phenylethenesulfonate
Molecular Weight (g/mol): 206.19
Molecular Formula: C8H7NaO3S
InChi Key: MNCGMVDMOKPCSQ-UHDJGPCESA-M
SMILES: [Na+].[O-]S(=O)(=O)\C=C\C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results