CAS RN 2040013-91-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-(2-Fluoro-6-nitrophenyl)homopiperazine, 97%, Thermo Scientific™
CAS: 2040013-91-8 Synonym: 1-(2-Fluoro-6-nitrophenyl)-1,4-diazepane