accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20440-95-3

IUPAC Name: 4-methyl-N-(4-methylphenyl)-N-phenylaniline
Synonyms: 4-methyl-N-(4-methylphenyl)-N-phenylaniline
Molecular Weight (g/mol): 273.38
Molecular Formula: C20H19N
InChi Key: YWKKLBATUCJUHI-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C)C=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results