accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(R)H3N(R)OHOOCH3CH3H3C

CAS RN 204587-91-7

IUPAC Name: (2R,3R)-4-(tert-butoxy)-3-hydroxy-4-oxobutan-2-aminium
Synonyms: (2R,3R)-4-(tert-butoxy)-3-hydroxy-4-oxobutan-2-aminium
Molecular Weight (g/mol): 176.24
Molecular Formula: C8H18NO3
InChi Key: KMUQLNSVLPPTEO-PHDIDXHHSA-O
SMILES: C[C@@H]([NH3+])[C@@H](O)C(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)