Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl (2R,3R)-3-amino-2-hydroxybutanoate 97%, Thermo Scientific™

Catalog No. AC338032500
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC338032500 250 mg
AC338031000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC338032500 Supplier Thermo Scientific Chemicals Supplier No. 338032500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 204587-91-7
Molecular Formula C8H17NO3
Molecular Weight (g/mol) 175.228
InChI Key KMUQLNSVLPPTEO-PHDIDXHHSA-N
Synonym tert-butyl 2r,3r-3-amino-2-hydroxybutanoate, butanoic acid,3-amino-2-hydroxy-, 1,1-dimethylethyl ester, 2r,3r, 2r,3r-tert-butyl 3-amino-2-hydroxybutanoate, tert-butyl 2>r,3r-3-amino-2-hydroxybutanoate, tert-butyl-2r,3r-3-amino-2-hydroxybutanoate
PubChem CID 2733830
IUPAC Name tert-butyl (2R,3R)-3-amino-2-hydroxybutanoate
SMILES CC(C(C(=O)OC(C)(C)C)O)N

Specifications

Quantity 250 mg
Formula Weight 175.23
Chemical Name or Material tert-Butyl (2R, 3R)-3-amino-2-hydroxybutanoate, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.