accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2049-55-0

IUPAC Name: triphenylphosphane boron
Synonyms: triphenylphosphine boron
Molecular Weight (g/mol): 273.1
Molecular Formula: C18H15BP
InChi Key: QFLQJPFCNMSTJZ-UHFFFAOYSA-N
SMILES: [B].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results