accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3H3COOH3C

CAS RN 2049-70-9

IUPAC Name: 1,3-diethyl 2-ethyl-2-methylpropanedioate
Synonyms: 1,3-diethyl 2-ethyl-2-methylpropanedioate
Molecular Weight (g/mol): 202.25
Molecular Formula: C10H18O4
InChi Key: ODRGILDUWDVBJX-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C)(CC)C(=O)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results