accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCl

CAS RN 2050-67-1

IUPAC Name: 3,3'-dichloro-1,1'-biphenyl
Synonyms: 3,3'-dichlorobiphenyl
Molecular Weight (g/mol): 223.1
Molecular Formula: C12H8Cl2
InChi Key: KTXUOWUHFLBZPW-UHFFFAOYSA-N
SMILES: ClC1=CC(=CC=C1)C1=CC(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results