accessibility menu, dialog, popup

UserName

Viewing profile as user:
Cl

CAS RN 2051-60-7

IUPAC Name: 2-chloro-1,1'-biphenyl
Synonyms: aroclor 1232 o-chlorodiphenyl 2-chlorodiphenyl
Molecular Weight (g/mol): 188.65
Molecular Formula: C12H9Cl
InChi Key: LAXBNTIAOJWAOP-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results