accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 205985-95-1

IUPAC Name: 2-methanesulfonylaniline
Synonyms: 2-methanesulfonylaniline
Molecular Weight (g/mol): 171.21
Molecular Formula: C7H9NO2S
InChi Key: AUSRSVRJOXYXMI-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=CC=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results