Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylsulphonylaniline hydrochloride, 95%

Catalog No. AC424840025
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424840025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC424840025 Supplier Thermo Scientific Chemicals Supplier No. 424840025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 205985-95-1 | C7H9NO2S | 171.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205985-95-1
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.21
MDL Number MFCD00216484
InChI Key AUSRSVRJOXYXMI-UHFFFAOYSA-N
Synonym 2-methylsulfonylaniline hydrochloride, 2-methylsulfonyl aniline hydrochloride, 2-methanesulfonylaniline hydrochloride, 2-methylsulphonylaniline hydrochloride, 2-methylsulphonyl aniline hydrochloride, 2-methylsulfonyl aniline, hcl, 2-methanesulfonyl-phenylammonium chloride, 2-methylsulfonyl phenylamine hydrochloride
PubChem CID 2735177
IUPAC Name 2-methylsulfonylaniline;hydrochloride
SMILES CS(=O)(=O)C1=CC=CC=C1N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 139°C to 141°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula SO2CH3C6H4NH2·HCl
Quantity 2.5 g
Solubility Information Solubility in water: slight
Formula Weight 207.68
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-Methylsulphonylaniline hydrochloride, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.