accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20601-83-6

IUPAC Name: selanylidenemercury
Synonyms: mercury selenide
Molecular Weight (g/mol): 279.56
Molecular Formula: HgSe
InChi Key: YQMLDSWXEQOSPP-UHFFFAOYSA-N
SMILES: [Se]=[Hg]
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)